Learn More
ORAI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA174181
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. In Western blot, this antibody detects a band at ∽49kDa. Rabbit anti Human ORAI1 antibody recognizes human ORAI1, also known as Transmembrane protein 142A (TMEM142A), a 32.
ORAI1 is a plasma membrane protein essential for store-operated calcium entry.Spezifikation
ORAI1 | |
Polyclonal | |
Unconjugated | |
Orai1 | |
calcium release-activated calcium channel 1; calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; CRACM1; D730049H07Rik; FLJ14466; hypothetical protein FLJ14466; hypothetical protein LOC402892; im:7146264; IMD9; ORAI calcium release-activated calcium modulator 1; ORAI calcium release-activated calcium modulator 1b; orai1; orai-1; orai1b; ORAT1; protein orai-1; RGD1311873; TAM2; TMEM142A; Transmembrane protein 142A; zFLJ14466; zgc:109721 | |
Rabbit | |
Affinity chromatography | |
RUO | |
109305, 304496, 84876 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5M848, Q8BWG9, Q96D31 | |
Orai1 | |
Peptide corresponding to a 16 residue sequence near the C-terminus of human ORAI1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |