Learn More
ORC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530109
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HepG2. Predicted reactivity: Mouse (91%), Rat (90%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. This gene encodes a subunit of the ORC complex. Several alternatively spliced transcript variants, some of which encode the same protein, have been reported for this gene.Spezifikation
ORC4 | |
Polyclonal | |
Unconjugated | |
ORC4 | |
HSORC4; mMmORC4; Orc4; ORC4L; Orc4P; Origin recognition complex subunit 4; origin recognition complex, subunit 4; origin recognition complex, subunit 4 homolog; origin recognition complex, subunit 4-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5000 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O43929 | |
ORC4 | |
Recombinant fragment corresponding to a region within amino acids 226 and 436 of Human ORC4L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |