Learn More
OX26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560345
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GREHAGCRFR FGRQDEGSEA TGFLPAAGEK TSGPLGNLAE ELNGYSRKKG GFSFR Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
OX26 Polyclonal antibody specifically detects OX26 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
OX26 | |
Polyclonal | |
Unconjugated | |
QRFP | |
26RFa; neuropeptide; Neuropeptide RF-amide; Orexigenic neuropeptide QRFP; P518; P518 precursor protein; P518 precursor protein (P518); prepro-QRFP; Pyroglutamylated arginine-phenylalanine-amide peptide; pyroglutamylated RFamide peptide; QRF-amide; Qrfp; RF(Arg-Phe)amide family 26 amino acid peptide (P518) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
347148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P83859 | |
QRFP | |
Recombinant protein corresponding to Human OX26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only