Learn More
DiscoverX Oxytocin
Click to view available options
Menge:
5 mg
Beschreibung
Oxytocin is a nine amino acid neurohypophysial hormone that acts as a neuromodulator in the brain. It is produced in the hypothalamus and secreted from the pituitary, and controls key parts of the reproductive system and brain, especially active during and after childbirth. Acts as an agonist for OXTR. Provided as 5mg of dry powder.
Spezifikation
Spezifikation
| Produkttyp | Control Ligands |
| Menge | 5 mg |