missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16427-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Two alternatively spliced transcript variants for this gene have been described. However, the full length nature of one of the variants has not been determined.Spezifikation
PAN2 | |
Polyclonal | |
Unconjugated | |
PAN2 | |
hPAN2, KIAA0710, PAN2, USP52 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103135, 408200, 9924 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q504Q3, Q6IE70, Q8BGF7 | |
Pan2 | |
PAN2 Fusion Protein Ag9670 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |