missing translation for 'onlineSavingsMsg'
Learn More

Parvalbumin Antibody (CL15070) [Unconjugated], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43649-100ul

 View more versions of this product

Product Code. 30279351

  • 605.00 € / 100 Mikroliter

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Description

Description

Parvalbumin Monoclonal antibody specifically detects Parvalbumin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Parvalbumin
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, Immunohistochemistry 1:2500-1:5000, Immunohistochemistry-Paraffin 1:2500-1:5000
D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV
This antibody was generated using a recombinant protein sequence of P20472, with the exact immunogen sequence remaining proprietary.
100 μL
Neuroscience
5816
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL15070
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.