Learn More
DiscoverX PathHunter™ U2OS INSRa Functional Assay
Artikelnummer. 17344219
Encompass
Change view
Click to view available options
Menge:
2 Fläschchen
Packungsgröße:
2 Ampullen
Product Code. Menge unitSize
17344219 2 Fläschchen 2 Ampullen
1 options
This item is not returnable. View return policy
Product Code. 17344219
Brand: DiscoverX 931008C3
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
This item is not returnable. View return policy
The PathHunter™ U2OS INSRa Functional Assay measures INSRa receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
The PathHunter™ U2OS INSRa Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated INSRa dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged INSRa and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Assay-Eingabe SH2 Recruitment
Zelllinie U2OS
Zur Verwendung mit (Anwendung) Cell Lines
Primärer Agonist Insulin, Human
Menge 2 Fläschchen
Zieleintrag INSRa
Zugriffsnummer NM_001079817