Learn More
DiscoverX PathHunter™ U2OS PRα Protein Interaction Cell Line
Click to view available options
Menge:
2 Fläschchen
Packungsgröße:
2 Ampullen
Beschreibung
The PathHunter™ PRα Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. testosterone, norgestrel) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged PRα NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of PRα, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Spezifikation
Spezifikation
| Assay-Eingabe | Protein Interaction |
| Zelllinie | U2OS |
| Zur Verwendung mit (Anwendung) | Cell Lines |
| Primärer Agonist | D(-)-Norgestrel |
| Menge | 2 Fläschchen |
| Zieleintrag | PRα |
| Zugriffsnummer | NM_001202474 |