Learn More
Intelsius™ PathoShield™
Artikelnummer. 17368599
Encompass_Preferred
Ansicht ändern
Click to view available options
Größe:
4
5
8
Dimensions (LxWxH):
191 x 145 x 22 mm
275 x 179 x 43 mm
291 x 191 x 102mm
Packungsgröße:
25 Stück
50 Stück
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Größe Dimensions (LxWxH) Packungsgröße
17368599 4 291 x 191 x 102mm 25 Stück
17378599 8 191 x 145 x 22 mm 50 Stück
15681632 5 275 x 179 x 43 mm 25 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17368599 Lieferant Intelsius™ Lieferanten-Nr. PA002
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
The PathoShield™ is a complete shipping solution designed for the transportation of Category A and Category B biological materials. Each system is pre-printed with the required markings and includes all the components required to transport your sample safely.
  • Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
  • Range of sizes suitable for shipping through postal and courier networks
  • Suitable for a wide range of primaries including blood vials, swabs and sample containers
  • Supplied flat packed for efficient storage and shipping prior to use
Spezifikation
Außenlänge (angloamerikanisch) 11.4 in.
Außenbreite (angloamerikanisch) 7.5 in.
Enthält Pouch
Größe 4
Außenhöhe (angloamerikanisch) 4.0 in.
Typ Sample Transport Packaging
Abmessungen (L x B) Pouch: 228 x 250 mm (8.9 x 9.8 in.)
Dimensions (LxWxH) 291 x 191 x 102mm