Learn More
PCK1 Mouse anti-Mouse, Unconjugated, Clone: 2F11B5, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66862-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The PCK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PCK1. This antibody recognizes mouse, rat, pig, human antigen. The PCK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| PCK1 | |
| Monoclonal | |
| 760 μg/mL | |
| Mouse | |
| Primary | |
| Mouse, Rat, Pig, Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2F11B5 | |
| Unconjugated | |
| 150 μL | |
| 5105 | |
| IgG1 |