missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16754-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Spezifikation
PCK1 | |
Polyclonal | |
Unconjugated | |
PCK1 | |
PCK1, PEPCK C, PEPCK1, PEPCKC, Phosphoenolpyruvate carboxykinase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18534, 362282, 5105 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07379, P35558, Q9Z2V4 | |
PCK1 | |
PCK1 Fusion Protein Ag10261 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |