missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PCM1 (Human) Recombinant Protein
Human PCM1 full-length ORF ( AAH00453, 1 a.a. - 530 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00005108-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MATGGGPFEDGMNDQDLPNWSNENVDDRLNNMDWGAQQKKANRSSEKNKKKFGVESDKRVTNDISPESSPGVGRRRTKTPHTFPHSRYMSQMSVPEQAELEKLKQRINFSDLDQRSIGSDSQGRATAANNKRQLSENRKPFNFLPMQINTNKSKDASTSPPNRETIGSAQCKELFASALSNDLLQNCQVSEEDGRGEPAMESSQIVSRLVQIRDYITKASSMREDLVEKNERSANVERLTHLIDHLKEQEKSYMKFLKKILARENEEEDVRTIDSAVGSGSVAESTSLNIDVQSEASDTTARDPQQEPMEEIENLKKQHDLLKRMLQQQEQLRALQGRQAALLALQHKAEQAIAVMDDSVVAETAGSLSGVSITSELNEELNDLIQRFHNQLRDSQPPAVPDNRRQAESLSLTREVSQSRKPSASERLPDEKVELFSKMRVLQEKKQKMDKLLGELHTLRDQHLNNSSSSPQRSVDQRSTSAPSASVGLAPVVNGESNSLTSSVPYPTASLVSQNESENEGHLNPSEKLQ
Spezifikation
AAH00453 | |
pericentriolar material 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCM1 | |
GST |
5108 | |
Wheat germ expression system | |
25 μg | |
PTC4 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |