missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L2 (PDCD1LG2) Mouse anti-Human, Clone: OTI8G8, lyophilized, TrueMAB™
Beschreibung
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD273 (PDCD1 Ligand 2, Programmed cell death 2 ligand 2, PD-L2, B7-DC) is involved in cell-mediated immune responses initiated by T lymphocytes that are stimulated by cognate peptides bound to MHC molecules on antigen-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as CD273 that mediate inhibitory signals from the APC (antigen presenting cell). CD273 can bind two different but related ligands, PDL-1 and PDL-2, both of which are thought act as a negative regulator of T cell activation. However, it has been suggested that PDL-2 can act to stimulate an immunogenic response through and alternative receptor from PD-1. At least three isoforms of PDL-2 are known to exist; CD273 is specific to the largest isoform. Diseases associated with CD273 include cysticercosis, and myasthenic syndrome.
Spezifikation
Spezifikation
| Antigen | PD-L2 (PDCD1LG2) |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI8G8 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 8% trehalose and no preservative; pH 7.3 |
| Gen | PDCD1LG2 |
| Gen-Zugriffsnummer | Q9BQ51 |
| Gen-Alias | B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2 |
| Gensymbole | PDCD1LG2 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?