Learn More
PDCD10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-47195-0.1MG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PDCD10 Polyclonal specifically detects PDCD10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Spezifikation
| PDCD10 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4) with No Preservative | |
| PDCD10 | |
| Recombinant human CCM3 | |
| 0.1 mg | |
| Primary | |
| The polyclonal antibody will detect recombinant human CCM-3 in WB and native CCM-3 in IHC | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:10-1:500 | |
| apoptosis-related protein 15, cerebral cavernous malformations 3, MGC1212, MGC24477, programmed cell death 10, programmed cell death protein 10, TF-1 cell apoptosis-related protein 15, TFAR15CCM3Cerebral cavernous malformations 3 protein | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11235 | |
| Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |