missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCD2 Rabbit anti-Human, Mouse, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10725-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
10725-1-AP is a rabbit polyclonal antibody raised against full-length PDCD2 of human origin.
Stem cells offer a therapeutic promise for many diseases. In the case of diabetes, stem cells offer the promise that beta cells may be generated ex vivo for the possible transplantation and cure of Type I diabetes (Couri CE, et al. ). For this to happen, stem cells will need to be isolated and expanded, they will need to be differentiated toward the beta cell lineage, and the differentiated cell progeny used for transplantation may need to be isolated from complex cell mixtures following differentiation. Appropriate markers targeting cell surface molecules on developing and mature pancreatic cells will be required for such research.Spezifikation
| PDCD2 | |
| Polyclonal | |
| Unconjugated | |
| PDCD2 | |
| PDCD2, programmed cell death 2, RP8, Zinc finger protein Rp 8, ZMYND7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18567, 393370, 5134 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P46718, Q16342 | |
| PDCD2 | |
| PDCD2 Fusion Protein Ag1139 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur