missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDCL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16057-1-AP-150UL
This item is not returnable.
View return policy
Description
Phosducin-like protein is a putative modulator of heterotrimeric G proteins. The protein shares extensive amino acid sequence homology with phosducin, a phosphoprotein expressed in retina and pineal gland. Both phosducin-like protein and phosphoducin have been shown to regulate G-protein signaling by binding to the beta-gamma subunits of G proteins.Specifications
| PDCL | |
| Polyclonal | |
| Unconjugated | |
| PDCL | |
| PDCL, PhLP, phosducin like, Phosducin like protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5082, 64013, 67466 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13371, Q63737, Q9DBX2 | |
| PDCL | |
| PDCL Fusion Protein Ag9029 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction