missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-9,10-dione, 98%, BLD Pharm™

Artikelnummer. 31448102
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31448102 10 g
31434532 100 g
31434415 1 g
31447066 5 g
31439114 25 g
31441653 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31448102 Lieferant BLD Pharm Lieferanten-Nr. BD10539510g
 Eingeschränkter Artikel
Verfügbar ab: 25-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material Phenanthrene-9,10-dione
CAS 84-11-7
Summenformel C14H8O2
MDL-Nummer MFCD00001163
Menge 10 g
SMILES O=C1C(=O)C2=CC=CC=C2C2=CC=CC=C12
Molekulargewicht (g/mol) 208.21
Reinheit (%) 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.