Learn More
Phospho-ATRIP (Ser239) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535392
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, non-human primate and sheep based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.
ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.Spezifikation
Phospho-ATRIP (Ser239) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 0.1mg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q8BMG1, Q8WXE1 | |
ATRIP | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser239 of human ATRIP conjugated to KLH | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ATRIP | |
6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
235610, 84126 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |