missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CtIP (Ser327) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA564481
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Phospho-CtIP (Ser327) Polyclonal Antibody detects endogenous levels of CtIP only when phosphorylated at Ser327.
Phospho-CtIP (Ser327) Polyclonal antibody specifically detects Phospho-CtIP (Ser327) in Human, Mouse samples. It is validated for Western Blot.Spezifikation
Phospho-CtIP (Ser327) | |
Polyclonal | |
Unconjugated | |
Rbbp8 | |
9930104E21Rik; COM1; CtBP-interacting protein; CtIP; DNA endonuclease RBBP8; JWDS; RB binding protein 8, endonuclease; RBBP8; RBBP-8; retinoblastoma binding protein 8; retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RGD1308872; RIM; SAE2; SCKL2; Sporulation in the absence of SPO11 protein 2 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
225182, 5932 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80YR6, Q99708 | |
Rbbp8 | |
A synthesized peptide derived from human CTIP (Phospho-Ser327) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |