missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-eIF4B (Ser422) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517615
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.Spezifikation
Phospho-eIF4B (Ser422) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P23588, Q8BGD9 | |
Eif4b | |
Synthetic phosphopeptide corresponding to residues surrounding pSer422 of human eIF4B | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Eif4b | |
2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1975, 300253, 75705 | |
-20°C | |
Liquid |