Learn More
Phospho-IRS1 (Ser312) Recombinant Polyclonal Antibody (18HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710778
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with Monkey, Bovine, Horse and Cat. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Phospho-IRS1 (Ser312) Recombinant Polyclonal antibody specifically detects Phospho-IRS1 (Ser312) in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Phospho-IRS1 (Ser312) | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P35568, P35569 | |
IRS1 | |
Phosphopeptide corresponding to Human IRS1 Ser312 (aa 309-318) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
18HCLC | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
Rabbit | |
Protein A | |
RUO | |
16367, 3667 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |