missing translation for 'onlineSavingsMsg'
Learn More

PI-103, MedChemExpress

Product Code. 16463288
Encompass_Preferred
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 Milligramm
5 Milligramm
50 Milligramm
This item is not returnable. View return policy

Product Code. 16463288

Brand: Medchem Express HY10115

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

MedChemExpress PI-103 is a potent PI3K and mTOR inhibitor with IC50s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α, p110β, p110δ, p110γ, mTORC1, and mTORC2. PI-103 also inhibits DNA-PK with an IC50 of 2 nM. PI-103 induces autophagy.

PI-103 is a potent PI3K and mTOR inhibitor with IC50s of 8 nM, 88 nM, 48 nM, 150 nM, 20 nM, and 83 nM for p110α, p110β, p110δ, p110γ, mTORC1, and mTORC2. PI-103 also inhibits DNA-PK with an IC50 of 2 nM. PI-103 induces autophagy.

Specifications

Chemischer Name oder Material PI-103
CAS 371935-74-9
Farbe Light Green
Hinweise zur Reinheitsqualität Research
Summenformel C19H16N4O3
Menge 10 mg
Löslichkeitsinformationen DMSO : 10 mg/mL (28.71 mM; Need ultrasonic)
SMILES OC1=CC(C2=NC3=C(C(N4CCOCC4)=N2)OC5=C3C=CC=N5)=CC=C1
Molekulargewicht (g/mol) 348.36
Formelmasse 348.36
Reinheit (%) 98.01%
Güte Research
Physikalische Form Powder
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.