missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIBF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14413-1-AP-150UL
Additional Details : Netto-Gewicht : 0.00010kg
Description
This gene encodes a protein that is induced by the steroid hormone progesterone and plays a role in the maintece of pregcy. The encoded protein regulates multiple facets of the immune system to promote normal pregcy including cytokine synthesis, natural killer (NK) cell activity, and arachidonic acid metabolism. Low serum levels of this protein have been associated with spontaneous pre-term labor in humans. This protein may promote the proliferation, migration and invasion of glioma.Specifications
PIBF1 | |
Polyclonal | |
Unconjugated | |
PIBF1 | |
C13orf24, CEP90, KIAA1008, PIBF, PIBF1, RP11 505F3.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10464, 306104, 52023 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WXW3 | |
Pibf1 | |
PIBF1 Fusion Protein Ag5755 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |