missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542879
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MEAMWLLCVA LAVLAWGFLW VWDSSERMKS REQGGRLGAE SRTLLVIAHP Sequence homology: Human: 100%
PIGL Polyclonal antibody specifically detects PIGL in Human samples. It is validated for Western Blot.Spezifikation
PIGL | |
Polyclonal | |
Unconjugated | |
PIGL | |
CHIME; N-acetylglucosaminylphosphatidylinositol deacetylase; N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase; phosphatidylinositol glycan anchor biosynthesis class L; phosphatidylinositol glycan anchor biosynthesis, class L; phosphatidylinositol glycan, class L; phosphatidylinositol-glycan biosynthesis class L protein; PIGL; PIG-L | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9487 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y2B2 | |
PIGL | |
Synthetic peptide directed towards the N-terminal of human PIGL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |