Learn More
PITRM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552339
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ELRSQQSKPQ DASCLPALKV SDIEPTIPVT ELDVVLTAGD IPVQYCAQPT NGMVYFRAFS SLNTLPEELR PYVPLFCSVL TKLGCGLLDY REQAQQIELK TGGMSASPHV LPDDSHMDTY EQGVLFSSLC LDRNLPDMMQ LWSEI Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.
PITRM1 Polyclonal antibody specifically detects PITRM1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PITRM1 | |
Polyclonal | |
Unconjugated | |
PITRM1 | |
2310012C15Rik; AA410010; hPreP; KIAA1104; Metalloprotease 1; metalloprotease 1 (pitrilysin family); mKIAA1104; MP1; MP-1; Ntup1; nuclear transplantation upregulated protein 1; pitrilysin metallepetidase 1; pitrilysin metallopeptidase 1; pitrilysin metalloprotease 1; pitrilysin metalloproteinase 1; Pitrm1; PREP; PreP peptidasome; presequence protease, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10531 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JRX3 | |
PITRM1 | |
Recombinant protein corresponding to Human PITRM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only