Learn More
PKC eta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544600
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: KKGHQLLDPY LTVSVDQVRV GQTSTKQKTN KPTYNEEFCA NVTDGGHLEL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 85%
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.Spezifikation
PKC eta | |
Polyclonal | |
Unconjugated | |
PRKCH | |
Protein kinase C, eta, MGC26269, MGC5363, PKC-L, PKCL, PRKCL, nPKC-eta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100051560, 100152637, 100736222, 18755, 490722, 518542, 5583, 81749 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P24723, P23298, Q64617 | |
PRKCH | |
Synthetic peptide directed towards the N-terminal of human PRKCH | |
100 μL | |
Primary | |
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rat, Zebrafish | |
Antibody | |
IgG |