Learn More
PKD1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554202
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SDDQERCLQA ACCLSFGGEL SVSTDKSWGL HLCSCSPPGG GLWVEVYANH VLLMSDGKCG CPWCALNGKA EDRESQSPSS SASRQKNIWK TTSEAALSVV NEKTQAVVNE KTQAPLDCDN SAD Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 26%.
This gene encodes a member of the polycystin protein family containing 11 transmembrane domains, a receptor for egg jelly (REJ) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. The encoded protein may play a role in the male reproductive system. Alternative splice variants have been described but their biological nature has not been determined.Spezifikation
PKD1L1 | |
Polyclonal | |
Unconjugated | |
PKD1L1 | |
AA444596; PC1-like 1 protein; PK1L1; Pkd1l1; polycystic kidney disease 1 like 1; polycystic kidney disease protein 1-like 1; polycystin 1 like 1, transient receptor potential channel interacting; polycystin-1L1; PRO19563; UNQ5785/PRO19563 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
168507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDX9 | |
PKD1L1 | |
Recombinant protein corresponding to Human PKD1L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only