Learn More
PKP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14062-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Plays a role as a regulator of Rho activity during cytokinesis. May play a role in junctional plaques.Spezifikation
| PKP4 | |
| Polyclonal | |
| Unconjugated | |
| PKP4 | |
| p0071, PKP4, plakophilin 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 227937, 295625, 8502 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q68FH0, Q99569 | |
| PKP4 | |
| PKP4 Fusion Protein Ag5113 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |