missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Roth™ Plastic Bottle Baskets
Marke: Carl Roth™ 0869.1
Weitere Details : Netto-Gewicht : 0.25000kg
Spezifikation
Bottle Basket | |
Ideal For Transporting Hazardous Liquids | |
1 Pack | |
235 x 235mm | |
235 mm |
Plastic Coated Steel Wire | |
4 | |
1 Pack | |
235 mm | |
130 mm |