missing translation for 'onlineSavingsMsg'
Learn More

Plastin-L Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Plastin-L Ab-1, Mouse Monoclonal Antibody.

Marke:  Epredia MS-1326-P1

Weitere Details : Netto-Gewicht : 1.00000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 12619566

  • 523.00 € / 500 Mikroliter
Verfügbar ab: 03-07-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosteronse in andrgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.

Host Species: Mouse

Clone: LPL4A.1

Isotype: IgG1

Species Reactivity: Human. Does not react with Mouse. Others not known.

Epitope: Headpiece region

Immunogen: Human recombinant L-plastin protein

Molecular Weight: 62kDa

Positive Control: Human polymorphonuclear neutrophils and monocytes, MCF-7, Jurkat cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence (Fixed in methanol or 3% paraformaldehyde)
  • Immunoprecipitation (Native)

Note:

USA: RUO; Int'l: RUO

Spezifikation

Spezifikation

Plastin-L Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Cytoskeletal Protein
Human
IgG1
Immunofluorescence, Immunoprecipitation, Western Blot
LPL4A.1
Unconjugated
Human recombinant L-plastin protein
RUO
Primary
Concentrated
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote