Learn More
Ibidi μ-Plate 24 Well Black ID 14 mm Uncoated
Click to view available options
:
μ-Plate
Beschreibung
- A black 24 well plate with flat and clear bottom for high throughput applications in cell-based assays
- well ID: 14 mm
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Spezifikation
Spezifikation
| Typ | Zellkultur-Objektträger |
| Wells | 24 |
| Farbe | Black |
| Durchmesser (metrisch) | 14 mm |