Learn More
Ibidi μ-Plate 384 Well Black Glass Bottom
Artikelnummer. 17834794
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
15
Packungsgröße:
15 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
17834794 15 15 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17834794 Lieferant Ibidi Lieferanten-Nr. 88407
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
μ-Plate 384 Well Black Glass Bottom is a black 384 well plate with square wells and a #1.5H glass coverslip bottom.
  • A black 384 well plate with square wells and a #1.5H glass coverslip bottom for high throughput screening using TIRF and super-resolution microscopy
  • bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
  • sterilized
Spezifikation
Sterilität Steril
Typ Microscope Slide
Farbe Black
Menge 15