Learn More
PLBD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553557
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GDWFSYDGSP RAQIFRRNQS LVQDMDSMVR LMRYNDFLHD PLSLCKACNP QPNGENAISA RSDLNPANGS YPFQALRQRS HGGIDVKVTS MSLARILSLL AASGPTWDQV PPFQWSTS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
PLBD2 Polyclonal antibody specifically detects PLBD2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
PLBD2 | |
Polyclonal | |
Unconjugated | |
PLBD2 | |
1300012G16Rik; 66.3 kDa protein; 76 kDa protein; AU019810; LAMA-like protein 2; Lamina ancestor homolog 2; mannose-6-phosphate protein associated protein p76; mannose-6-phosphate protein p76; p76; phospholipase B domain containing 2; phospholipase B domain-containing protein 2; phospholipase B-like 2 32 kDa form; phospholipase B-like 2 45 kDa form; PLB homolog 2; Plbd2; Putative phospholipase B-like 2; Putative phospholipase B-like 2 15 kDa form; Putative phospholipase B-like 2 28 kDa form; Putative phospholipase B-like 2 32 kDa form; Putative phospholipase B-like 2 40 kDa form; Putative phospholipase B-like 2 45 kDa form; RDCR-0918-3; RDCR-0918-3 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
196463 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NHP8 | |
PLBD2 | |
Recombinant protein corresponding to Human PLBD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only