Learn More
PLCXD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563216
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DFNHFYAMDE THHKCLVLRI QEAFGNKLCP ACSVESLTLR TLWEKNCQVL IFYHCPFYKQ YPFLWPGKKI PAPWANTTS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
PLC-XD2 (PI-PLC X domain-containing protein 2) is a 305 amino acid protein that contains a domain that is present in many PLC isozymes, the PI-PLC X (X-box)domain. Both the X-box domain and the Y-box domain are important for catalytic activity in PLC proteins. X-box domains are conserved from prokaryotes to mammals. The gene encoding PLC-XD2 maps to human chromosome 3, which is made up of about 214 million bases encoding over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. There are two isoforms of PLC-XD2 that are produced as a result of alternative splicing events.Spezifikation
PLCXD2 | |
Polyclonal | |
Unconjugated | |
PLCXD2 | |
AW494639; EG433022; phosphatidylinositol specific phospholipase C X domain containing 2; phosphatidylinositol-specific phospholipase C, X domain containing 2; PI-PLC X domain-containing protein 2; PLCXD2; RGD1563504 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
257068 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0VAA5 | |
PLCXD2 | |
Recombinant protein corresponding to Human PLCXD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only