Learn More
Motic™ PM-Series Microscope
Petrographic that microscope can be used as a standard bright field microscope or as a polarizing microscope. Motic™ PM-Series Microscope is well suited for investigations in geology and mineralogy
Marke:  Motic™ 1101001703271
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11722348
  • 143.00 € / 1 Stück
Verfügbar ab: 23-07-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • The PM Series contains different models with Swing in/out analyzer & polarizer in monocular, binocular and trinocular version, or with 1/4 λ plate, full λ plate, Bertrand lens and a coaxial focusing system.
  • DIN EA achromatic objectives 4X, 10X and 40X.
  • Analyzer and polarizer.
  • Basic and advanced version available.