Learn More
Motic™ PM-Series Microscope
Petrographic that microscope can be used as a standard bright field microscope or as a polarizing microscope. Motic™ PM-Series Microscope is well suited for investigations in geology and mineralogy
Marke: Motic™ 1101001703271
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
- The PM Series contains different models with Swing in/out analyzer & polarizer in monocular, binocular and trinocular version, or with 1/4 λ plate, full λ plate, Bertrand lens and a coaxial focusing system.
- DIN EA achromatic objectives 4X, 10X and 40X.
- Analyzer and polarizer.
- Basic and advanced version available.