missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA565742
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Immunogen sequence: QGQGLRARGVP AFTDTTLDEP VPDDRY Highest antigen sequence identity to the following orthologs - mouse 67%, rat 37%.
PMAT(Slc29a4) is a member of the SLC29 family and encodes a plasma membrane protein with 11 transmembrane helices. This protein catalyzes the re-uptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It has been shown to transport several compounds, including serotonin, dopamine, and the neurotoxin 1-methyl-4-phenylpyridinium.Spezifikation
PMAT | |
Polyclonal | |
Unconjugated | |
SLC29A4 | |
ENT4; Equilibrative nucleoside transporter 4; FLJ34923; hENT4; mPMAT; Plasma membrane monoamine transporter; PMAT; PSEC0113; SLC29A4; solute carrier family 29 (equilibrative nucleoside transporter), member 4; solute carrier family 29 (nucleoside transporters), member 4; solute carrier family 29 member 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222962 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTT9 | |
SLC29A4 | |
Recombinant Human PMAT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only