missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMP22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567643
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with Mouse and Rat samples.
Peripheral Myelin Protein 22 (PMP22) is a 160 amino acid glycoprotein which is a member of the claudin family and is thought to play an important role in the structure of peripheral nerve myelin. Irregularities in its gene results in hereditary demyelinating neuropathies. PMP22 protein is present in the compact myelin of adult human peripheral nerve and its distribution mimics that of Myelin Protein zero (P0).Spezifikation
PMP22 | |
Polyclonal | |
Unconjugated | |
PMP22 | |
22kDa; Cd25; CMT1A; CMT1E; DSS; GAS3; GAS-3; Growth arrest-specific protein 3; HMSNIA; HNPP; peripheral myelin; peripheral myelin protein 22; peripheral myelin protein 22 kDa; PMP22; PMP-22; Protein CD25; SAG; schwann cell membrane glycoprotein; Sp110; SR13 myelin protein; Tr; trembler | |
Rabbit | |
affinity chromatography | |
RUO | |
18858, 24660, 5376 | |
-20°C | |
Liquid |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P16646, P25094, Q01453 | |
PMP22 | |
Synthesized peptide derived from human PMP22. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |