missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Polyethylen-Etiketten für Gefahrstoffe

Artikelnummer. p-8006280
Encompass_Preferred
Click to view available options
Angezeigter Text:
GefahrVerbrennungs-und Verätzungsgefahr
Korrosionspiktogramm/Achtung/Warning/Attention
Unit Size:
250 Stück
This item is not returnable. View return policy

Product Code. 10553258

Brand: Carl Roth™ LN55.1

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made of high glossy PE-foil. Carl Roth™ Polyethylene Hazardous Substances Labels feature signal words in German, English, and French.

  • High-glossy PE-foil
  • 100 μm thick
  • Permanent adhesive
  • Waterproof
  • Temperature stable from -20 to +80°C 
  • Signal words trilingual: German, English, French
  • 250 per roll in practical dispenser box

Specifications

Ausführung Glanz
Material Polyethylen-Folie
Beständig gegen Wasserfest und temperaturbeständig
Höhe (metrisch) 30 mm
Dicke 100 μ m
Breite (metrisch) 22 mm
Temperaturbereich (metrisch) -20 °C bis 80 °C
Form Rechteckig
Sprachen Deutsch, Englisch, Französisch
Angezeigter Text GefahrVerbrennungs-und Verätzungsgefahr
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.