missing translation for 'onlineSavingsMsg'
Learn More

Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, 99%, BLD Pharm™

Artikelnummer. 30963654
Encompass_Preferred
Ansicht ändern
Click to view available options
Menge:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30963654 1 kg
30953233 100 g
30963056 500 g
30956953 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30963654 Lieferant BLD Pharm Lieferanten-Nr. BD1487821000g
 Eingeschränkter Artikel
Verfügbar ab: 27-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemischer Name oder Material Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
CAS 299-27-4
Summenformel C6H11KO7
MDL-Nummer MFCD00064211
Menge 1 kg
SMILES O=C([O-])[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.[K+]
Molekulargewicht (g/mol) 234.25
Reinheit (%) 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.