missing translation for 'onlineSavingsMsg'
Learn More

Kalium-4-(1-pyrrolidinylcarbonyl)-phenyltrifluorborat, 95 %, Thermo Scientific™

Catalog Number p-7034226
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1983205-51-1
C11H12BF3KNO
281.127
MFCD16293904
HEBRRMDWGHQMOE-UHFFFAOYSA-N
potassium 4-pyrrolidine-1-carbonyl phenyl trifluoroborate, potassium 4-1-pyrrolidinylcarbonyl phenyltrifluoroborate, potassium trifluoro 4-pyrrolidine-1-carbonyl phenyl boranuide, amtb215, potassium 4-pyrrolidine-1-carbonyl-phenyl trifluoroborate
73995281
Kalium;Trifluor-[4-(Pyrrolidin-1-Carbonyl)Phenyl]Boranuid
[B-](C1=CC=C(C=C1)C(=O)N2CCCC2)(F)(F)F.[K+]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1983205-51-1
C11H12BF3KNO
281.127
MFCD16293904
HEBRRMDWGHQMOE-UHFFFAOYSA-N
potassium 4-pyrrolidine-1-carbonyl phenyl trifluoroborate, potassium 4-1-pyrrolidinylcarbonyl phenyltrifluoroborate, potassium trifluoro 4-pyrrolidine-1-carbonyl phenyl boranuide, amtb215, potassium 4-pyrrolidine-1-carbonyl-phenyl trifluoroborate
73995281
Kalium;Trifluor-[4-(Pyrrolidin-1-Carbonyl)Phenyl]Boranuid
[B-](C1=CC=C(C=C1)C(=O)N2CCCC2)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Potassium 4-(1-pyrrolidinylcarbonyl)phenyltrifluoroborate
Menge 1 g
Formelmasse 281.13
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'