missing translation for 'onlineSavingsMsg'
Learn More

Kalium-4-(Benzylaminocarbonyl)phenyltrifluorborat, 95 %, Thermo Scientific™

Click to view available options
Menge:
1g
5g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2017555-07-4
C14H12BF3KNO
317.16
MFCD16293900
RDEFKKXMPRJFFH-UHFFFAOYSA-N
potassium 4-benzylamino-1-carbonyl phenyl trifluoroborate, potassium 4-benzylcarbamoyl phenyl trifluoroboranuide, amtb218, c14h12bf3no.k, potassium 4-benzylamino-1-carbonyl-phenyl trifluoroborate
71306520
Kalium;[4-(Benzylcarbamoyl)phenyl]-trifluoroboranuid
[B-](C1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2)(F)(F)F.[K+]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2017555-07-4
C14H12BF3KNO
317.16
MFCD16293900
RDEFKKXMPRJFFH-UHFFFAOYSA-N
potassium 4-benzylamino-1-carbonyl phenyl trifluoroborate, potassium 4-benzylcarbamoyl phenyl trifluoroboranuide, amtb218, c14h12bf3no.k, potassium 4-benzylamino-1-carbonyl-phenyl trifluoroborate
71306520
Kalium;[4-(Benzylcarbamoyl)phenyl]-trifluoroboranuid
[B-](C1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemischer Name oder Material Potassium 4-(benzylaminocarbonyl)phenyltrifluoroborate
Menge 1g
Formelmasse 317.16
Reinheit (%) 95%