missing translation for 'onlineSavingsMsg'
Learn More
Learn More
proBDNF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSB00015W
Weitere Details : Netto-Gewicht : 0.99000kg
Beschreibung
Glycerol (1:1) may be added for additional stability. Reconstitute with 100 μL of sterile water.
plays a role in the development of hippocampal long term potentiation; involved in regulation of synaptic plasticity.Spezifikation
proBDNF | |
Polyclonal | |
Bdnf | |
Brain-derived neurotrophic factor precursor, proBDNF, pro BDNF | |
Rabbit | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P21237, P23363 | |
BDNF | |
A synthetic peptide from mouse proBDNF | |
RUO | |
12064, 24225 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |