Learn More
DiscoverX Prolactin
Click to view available options
Menge:
10 μg
Beschreibung
Prolactin is a pituitary hormone that promotes lactation, commonly referred to as the milk hormone. Also has a wide range of regulatory functions on the reproductive and immune system, such as growth, differentiation, anti-apoptotic factors, and influences on hematopoiesis, angiogenesis, and blood clotting. Acts as an agonist for PRLR. Provided as 10μg of dry powder alongside reconstitution buffer.
Spezifikation
Spezifikation
| Produkttyp | Control Ligands |
| Menge | 10 μg |