missing translation for 'onlineSavingsMsg'
Learn More

Propyl Acetate, ≥99.5%, Honeywell™

Catalog Number p-7120732
Click to view available options
Menge:
1 L
2.5 L
Verpackung:
Glass Bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
109-60-4
C5H10O2
102.13
MFCD00009372
YKYONYBAUNKHLG-UHFFFAOYSA-N
n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
7997
CHEBI:40116
propyl acetate
CCCOC(C)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
109-60-4
C5H10O2
102.13
MFCD00009372
YKYONYBAUNKHLG-UHFFFAOYSA-N
n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
7997
CHEBI:40116
propyl acetate
CCCOC(C)=O

≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Propyl acetate
Schmelzpunkt -95°C
Farbe Colorless
Dichte 0.888g/mL at 25°C
Siedepunkt 102°C
Flammpunkt 14°C (57.2°F)
Zündpunkt 842°F
Verpackung Glass Bottle
Strukturformel CH3COOCH2CH2CH3
Menge 1 L
UN-Nummer UN1276
Beilstein 1740764
Dampfdichte 3.5 (vs air)
Dampfdruck 25mmHg (20°C)
Formelmasse 102.13g/mol
Reinheit (%) ≥99.5%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'