Learn More
Thermo Scientific Chemicals Proteasom-Inhibitor II
Marke: Thermo Scientific™ J64973.#0
Weitere Details : Netto-Gewicht : 0.00005kg
Beschreibung
Potent and cell permeable proteasome inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Weiß bis cremefarben | |
C29H39N3O5 | |
Z-LLF-CHO; Z-Leu-Leu-Phe-CHO | |
Pulver | |
5 mg |
Inhibitor | |
509.64 | |
-30°C to -10°C | |
Potenter und zelldurchlässiger Proteasom-Inhibitor |
RUO – Research Use Only