Learn More
Eppendorf™ Protection Filter Set
Designed for use with Eppendorf Reference™ 2, Eppendorf Research™ plus and Eppendorf Xplorer™ (plus) pipettes
Marke: Eppendorf™ 4920622009
Weitere Details : Netto-Gewicht : 0.01100kg
Beschreibung
- Available in a variety of colors
- Protection Filter Sets include 10 filters and 1 filter sleeve
- For fixed- and variable-volume pipettes