missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Schutzanzug 4565
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4XL
Förpackningsstorlek
20 Stück
Beskrivning
- Wear 3M™ Protective Coverall 4565 for an ideal barrier to hazardous particles, limited liquid splashes and sprays to Type 4
- Fresh white coverall 4565 has recognizable red taped seams and meets a variety of standards, including chemical protective clothing EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1, and Type 4 Liquid Spray EN 14605
- Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles
- Offers Til class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold out as well as liquid spray resistance
- Made with a non-microporous PE laminate for chemical protection with permeation-tested resistance against certain chemicals
- Knitted cuffs, elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination
- Anti-static treatment meeting EN 1149, which reduces static discharge and risk of spark and ignition
Specifikationer
Specifikationer
| Produkttyp | Reinraumoveralls |
| Größe | X-Large |
| Geschlecht | Unisex |
| Material | Polypropylen |
| Farbe | White |
| Einwegprodukt | Single-use |
| Taillenform | Elastisch |
| Knöchelform | Elastisch |
| Stulpenform | Knit |
| Flaschenhalstyp | Hooded |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering