Learn More
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask
Marke: Pyrex 1080/02M
Weitere Details : Netto-Gewicht : 0.47000kg
Beschreibung
- Complies with ISO 24450
- With wide neck to facilitate easy addition of solid samples etc
- Flat base for standing on benches and hotplates etc
- Manufactured from Pyrex™ glass for high resistance to thermal shock
- Excellent chemical durability for contamination free performance