Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Keep material securely dry. QuickFitTM Borosilicate glass drying angled cone to fit QuickFitTTM apparatus.
Marke: Quickfit MF23/4
Weitere Details : Netto-Gewicht : 0.08000kg
Beschreibung
- For use with a desiccant such as calcium chloride
- Ground glass cone to fit Quickfit™ apparatus