Learn More
Quickfit™ Flanschdeckel aus Borosilikatglas für Quickfit™ Weithalskolben
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid is comprised of precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid includes one or more sockets to accept other Quickfit™ components.
Marke: Quickfit MAF5/2
Weitere Details : Netto-Gewicht : 0.53000kg
Beschreibung
- With precision ground flat-flange joints
- Fits Quickfit™ wide neck reaction flasks and culture vessels
- Each lid with one or more sockets to accept other Quickfit™ components